Imprimer
Anti-Retinol binding protein 4 Mouse Monoclonal Antibody
Fournisseur: Biorbyt
BIRBORB27759-100EA 526 CHF
BIRBORB27759-100
Anti-Retinol binding protein 4 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Retinol binding protein 4
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human