Imprimer

Anti-FABP4 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27754-100EA 0 CHF
BIRBORB27754-100
Anti-FABP4 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR