Imprimer

Anti-ADIPOQ Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27751-100EA 526 CHF
BIRBORB27751-100
Anti-ADIPOQ Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR