Imprimer

Anti-IgG Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27720-1EA 0 CHF
BIRBORB27720-1 BIRBORB27720-0.5
Anti-IgG Goat Polyclonal Antibody
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR