Imprimer

Anti-IgM Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27698-0.5EA 0 CHF
BIRBORB27698-0.5 BIRBORB27698-0.25
Anti-IgM Goat Polyclonal Antibody
Anticorps
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR