Imprimer

Anti-FGFR1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27682-100EA 0 CHF
BIRBORB27682-100
Anti-FGFR1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: FGFR1 (fibroblast growth factor receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR