Imprimer

Anti-BAK1 Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27661-100EA 0 CHF
BIRBORB27661-100
Anti-BAK1 Rabbit polyclonal antibody
Anticorps
Type: Primary
Antigen: BAK1 (BCL2-antagonist/killer 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR