Imprimer

Anti-RETN Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27655-100EA 589 CHF
BIRBORB27655-100
Anti-RETN Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: RETN (resistin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR