Imprimer

Anti-IL2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27653-100EA 0 CHF
BIRBORB27653-100
Anti-IL2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: IL2 (interleukin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR