Imprimer

Anti-FADD Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27648-100EA 515 CHF
BIRBORB27648-100
Anti-FADD Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR