Imprimer

Anti-LGALS1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27631-100EA 589 CHF
BIRBORB27631-100
Anti-LGALS1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: LGALS1 (lectin, galactoside-binding, soluble, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR