Imprimer

Anti-APOD Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27600-100EA 0 CHF
BIRBORB27600-100
Anti-APOD Rabbit polyclonal antibody
Anticorps
Type: Primary
Antigen: APOD (apolipoprotein D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR