Imprimer

Anti-MMP10 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27592-100EA 616 CHF
BIRBORB27592-100
Anti-MMP10 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MMP10 (matrix metallopeptidase 10 (stromelysin 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR