Imprimer

Anti-MDM2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27590-100EA 515 CHF
BIRBORB27590-100
Anti-MDM2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR