Imprimer

Anti-HSD11B1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27584-100EA 0 CHF
BIRBORB27584-100
Anti-HSD11B1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HSD11B1 (hydroxysteroid (11-beta) dehydrogenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR