Imprimer

Anti-TGFA Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27573-100EA 0 CHF
BIRBORB27573-100
Anti-TGFA Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR