Imprimer
Anti-PSEN2 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
BIRBORB27571-100EA 0 CHF
BIRBORB27571-100
Anti-PSEN2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now