Imprimer

Anti-HDAC1 Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27563-100EA 424 CHF
BIRBORB27563-100
Anti-HDAC1 Rabbit polyclonal antibody
Anticorps
Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR