Imprimer

Anti-PIN-Cocktail 1 P504S / p63 None Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27517-5EA 640 CHF
BIRBORB27517-5
Anti-PIN-Cocktail 1 P504S / p63 None Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PIN-Cocktail 1 P504S / p63
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG+IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR