Imprimer

Anti-Pentosidine Mouse Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB27503-50EA 0 CHF
BIRBORB27503-50
Anti-Pentosidine Mouse Monoclonal Antibody (Biotin)
Anticorps
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: Pentosidine
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR