Imprimer

Anti-Borrelia Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27489-200EA 774 CHF
BIRBORB27489-200
Anti-Borrelia Rabbit polyclonal antibody
Anticorps
Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Borrelia
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR