Imprimer

Anti-Mammaglobin Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27427-5EA 0 CHF
BIRBORB27427-5
Anti-Mammaglobin Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Mammaglobin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR