Imprimer

Anti-Fibronectin Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27333-0.5EA 0 CHF
BIRBORB27333-0.5
Anti-Fibronectin Rabbit polyclonal antibody
Anticorps
ImmunofluorescenceA 1:40; Immunochemistry: 1:200 - 1:600

Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR