Imprimer

Anti-COL9 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27327-0.5EA 0 CHF
BIRBORB27327-0.5
Anti-COL9 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: COL9 (collagen, type IX)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR