Imprimer

Anti-COL5 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27325-0.5EA 0 CHF
BIRBORB27325-0.5
Anti-COL5 Rabbit Polyclonal Antibody
Anticorps
ImmunofluorescenceA 1:40; Immunochemistry: 1:250; ELISA: > 1:100-1:200

Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR