Imprimer

Anti-COL2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27312-0.5EA 0 CHF
BIRBORB27312-0.5 BIRBORB27312-0.1
Anti-COL2 Rabbit Polyclonal Antibody
Anticorps
ImmunofluorescenceA 1:40; Immunochemistry: 1:100 - 1:500

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR