Imprimer

Anti-COL1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27308-0.1EA 0 CHF
BIRBORB27308-0.1 BIRBORB27308-0.5
Anti-COL1 Rabbit Polyclonal Antibody
Anticorps
ImmunofluorescenceA 1:40; Immunochemistry: 1:500

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR