Imprimer

Anti-COL1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27304-0.5EA 0 CHF
BIRBORB27304-0.5 BIRBORB27304-0.1
Anti-COL1 Rabbit Polyclonal Antibody
Anticorps
ImmunofluorescenceA 1:80; Immunochemistry: 1:1000; ELISA: >1:200

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR