Imprimer

Anti-alpha Defensin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27295-100EA 1174 CHF
BIRBORB27295-100
Anti-alpha Defensin Mouse Monoclonal Antibody
Anticorps
Immunochemistry: 0.2-0.5 ?g/ml

Type: Primary
Antigen: alpha Defensin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR