Imprimer

Anti-FIGF Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27289-200EA 0 CHF
BIRBORB27289-200
Anti-FIGF Rabbit polyclonal antibody
Anticorps
Type: Primary
Antigen: FIGF (c-fos induced growth factor (vascular endothelial growth factor D))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR