Imprimer

Anti-VEGFA Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB27288-100EA 0 CHF
BIRBORB27288-100
Anti-VEGFA Rabbit Polyclonal Antibody
Anticorps
Western Blot: 5-10 ?g/ml; ELISA: 0.5-2 ?g/ml

Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR