Imprimer

Anti-DEFB1 Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27279-100EA 0 CHF
BIRBORB27279-100
Anti-DEFB1 Rabbit polyclonal antibody
Anticorps
Western Blot: 1-10 ?g/ml; 0.5-1 ?g/ml ELISA; Immunochemistry: 2-10 ?g/ml (not tested)

Type: Primary
Antigen: DEFB1 (defensin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR