Imprimer

Anti-DEFB4A Rabbit polyclonal antibody

Fournisseur: Biorbyt
BIRBORB27276-100EA 0 CHF
BIRBORB27276-100
Anti-DEFB4A Rabbit polyclonal antibody
Anticorps
Western Blot: 1:1000 - 1:5000; ELISA: 1:10000 - 1:50000

Type: Primary
Antigen: DEFB4A (defensin, beta 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR