Imprimer

Anti-CDH1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27241-5EA 0 CHF
BIRBORB27241-5
Anti-CDH1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR