Imprimer

Anti-Cytokeratin (High Molecular Weight) 34betaE12 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27208-5EA 0 CHF
BIRBORB27208-5
Anti-Cytokeratin (High Molecular Weight) 34betaE12 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Cytokeratin (High Molecular Weight) 34betaE12
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR