Imprimer

Anti-KRT14 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27203-5EA 504 CHF
BIRBORB27203-5
Anti-KRT14 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: KRT14 (keratin 14)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR