Imprimer

Anti-IgG Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26912-5EA 539 CHF
BIRBORB26912-5
Anti-IgG Goat Polyclonal Antibody
Anticorps
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR