Imprimer
Anti-Procollagen Type III Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
BIRBORB26854-100EA 0 CHF
BIRBORB26854-100
Anti-Procollagen Type III Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:100-1:200; IRA: 1:100-1:200; ELISA: 1:100-1:200; Immunochemistry: 1:500
Type: Primary
Antigen: Procollagen Type III
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human