Imprimer

Anti-COL4 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26850-0.5EA 0 CHF
BIRBORB26850-0.5
Anti-COL4 Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:80; Immunochemistry: 1:500; ELISA: 1:200

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR