Imprimer

Anti-COL2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26845-0.5EA 1280 CHF
BIRBORB26845-0.5
Anti-COL2 Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:40

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR