Imprimer

Anti-COL5 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26841-0.5EA 0 CHF
BIRBORB26841-0.5
Anti-COL5 Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:40; Immunochemistry: 1:500; ELISA: 1:200

Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR