Imprimer

Anti-SPEA Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26802-1EA 1280 CHF
BIRBORB26802-1
Anti-SPEA Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SPEA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR