Imprimer

Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26749-100EA 0 CHF
BIRBORB26749-100
Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR