Imprimer

Anti-Opioid Receptor, kappa Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26733-100EA 0 CHF
BIRBORB26733-100
Anti-Opioid Receptor, kappa Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 1:5000; Western Blot: 1:1500

Type: Primary
Antigen: Opioid Receptor, kappa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR