Imprimer

Anti-Endorphin, beta Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26711-100EA 0 CHF
BIRBORB26711-100
Anti-Endorphin, beta Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Endorphin, beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR