Imprimer

Anti-Nicotinic beta 3 Rec. Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26705-100EA 0 CHF
BIRBORB26705-100
Anti-Nicotinic beta 3 Rec. Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 1:50; Western Blot: 1:100

Type: Primary
Antigen: Nicotinic beta 3 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR