Imprimer

Anti-UB Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26678-0.2EA 598 CHF
BIRBORB26678-0.2
Anti-UB Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:5000; Immunochemistry: 1:200; Western Blot: 1:1000

Type: Primary
Antigen: UB (ubiquitin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR