Imprimer

Anti-PRNP Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26666-100EA 0 CHF
BIRBORB26666-100
Anti-PRNP Mouse Monoclonal Antibody
Anticorps
ELISA: 1:20000-1:50000; Western Blot: 1:10000-1:25000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: PRNP (prion protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR