Imprimer

Anti-NR2A subunit Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB26651-50EA 352 CHF
BIRBORB26651-50
Anti-NR2A subunit Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: NR2A subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR