Imprimer

Anti-BSG Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26597-0.2EA 1362 CHF
BIRBORB26597-0.2
Anti-BSG Mouse monoclonal antibody
Anticorps
Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR